Please login to your online account to display your discounted pricing

2,5-Dichloronitrobenzene, 99%, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  209212500

 View more versions of this product

Catalog No. AC209212500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Flakes
Formula Weight 192
Quantity 250g
Color Yellow
EINECS Number 201-923-3
TSCA TSCA
RTECS Number CZ5260000
Melting Point 52° to 56°C
CAS Max % 100.0
Chemical Name or Material 2, 5-Dichloronitrobenzene
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
MDL Number MFCD00007074
Packaging Plastic bottle
CAS 89-61-2
Synonym 1, 4-Dichloro-2-nitrobenzene
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 5-Dichloronitrobenzene
Name Note 99%
CAS 89-61-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
Formula Weight 192
MDL Number MFCD00007074
Synonym 1, 4-Dichloro-2-nitrobenzene
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents