Please login to your online account to display your discounted pricing

2,5-Dichloronitrobenzene, 99%, ACROS Organics™

$54.80 - $54.80

Chemical Identifiers

Chemical Name or Material 2, 5-Dichloronitrobenzene
Name Note 99%
CAS 89-61-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
Formula Weight 192
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209212500

  SDS
thermo fisher scientific
209212500
Plastic bottle 250g Each for $54.80

AC209210050

  SDS
thermo fisher scientific
AC209210050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Flakes
Formula Weight 192
Quantity 250g
Color Yellow
EINECS Number 201-923-3
TSCA TSCA
RTECS Number CZ5260000
Melting Point 52° to 56°C
CAS Max % 100.0
Chemical Name or Material 2, 5-Dichloronitrobenzene
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
MDL Number MFCD00007074
Packaging Plastic bottle
CAS 89-61-2
Synonym 1, 4-Dichloro-2-nitrobenzene
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.