Please login to your online account to display your discounted pricing

2,5-Dichloronitrobenzene, 99%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC209210050

 View more versions of this product

Catalog No. AC209210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Flakes
Assay ≥98.5%
Formula Weight 192
Quantity 5g
Color Yellow
Packaging Glass Bottle
TSCA TSCA
Melting Point 52° to 56°C
RTECS Number CZ5260000
EINECS Number 201-923-3
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Synonym 1, 4-Dichloro-2-nitrobenzene
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Chemical Name or Material 2, 5-Dichloronitrobenzene
Linear Formula Cl2C6H3NO2
MDL Number MFCD00007074
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
CAS 89-61-2
Name Note 99%

Specifications

Chemical Name or Material 2, 5-Dichloronitrobenzene
Name Note 99%
CAS 89-61-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H3Cl2NO2
Linear Formula Cl2C6H3NO2
Formula Weight 192
MDL Number MFCD00007074
Synonym 1, 4-Dichloro-2-nitrobenzene
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents