missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorohydroquinone, 97%

$54.46 - $181.00

Chemical Identifiers

CAS 824-69-1
Molecular Formula C6H4Cl2O2
Formula Weight 179
MDL Number MFCD00041749
Synonym 1,4-Dichloro-2,5-dihydroxybenzene
Chemical Name or Material 2,5-Dichlorohydroquinone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2128503 SDS Alfa Aesar™
B2128503
1g Each for $54.46
AAB2128506 SDS Alfa Aesar™
B2128506
5g Each for $181.00
Specifications Description & Specifications

Chemical Identifiers

824-69-1
179
1,4-Dichloro-2,5-dihydroxybenzene
C6H4Cl2O2
MFCD00041749
2,5-Dichlorohydroquinone

Specifications

168° to 171°C
824-69-1
179
1,4-Dichloro-2,5-dihydroxybenzene
97%
1g
C6H4Cl2O2
MFCD00041749
2,5-Dichlorohydroquinone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA