2,5-Dichlorobenzyl bromide, 97%, Acros Organics™
Manufacturer:  Acros Organics AC443690050
 View more versions of this product
Catalog No. AC443690050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
2,5-Dichlorobenzyl bromide
134.0°C to 138.0°C (14.0 Torr)
5g
C7H5BrCl2
UUVDOPTUDWJHFK-UHFFFAOYSA-N
2-(bromomethyl)-1,4-dichlorobenzene
3495744
97%
Authentic
41.0°C to 43.0°C
85482-13-9
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
C1=CC(=C(C=C1Cl)CBr)Cl
239.921
239.93
Chemical Identifiers
85482-13-9
239.921
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
2-(bromomethyl)-1,4-dichlorobenzene
C7H5BrCl2
UUVDOPTUDWJHFK-UHFFFAOYSA-N
3495744
C1=CC(=C(C=C1Cl)CBr)Cl
Safety and Handling
Safety and Handling
GHS H Statement Causes severe skin burns and eye damage.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.
Danger
Documents