missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzyl bromide, 97%

Manufacturer:  Alfa Aesar™ L1377503

 View more versions of this product

Catalog No. AAL1377503



Specifications

Specifications

2,5-Dichlorobenzyl bromide
1g
C7H5BrCl2
UN3261
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
C1=CC(=C(C=C1Cl)CBr)Cl
239.921
239.93
39°C to 43°C
85482-13-9
MFCD00236034
637803
UUVDOPTUDWJHFK-UHFFFAOYSA-N
2-(bromomethyl)-1,4-dichlorobenzene
3495744
97%

Chemical Identifiers

85482-13-9
239.921
UUVDOPTUDWJHFK-UHFFFAOYSA-N
3495744
C1=CC(=C(C=C1Cl)CBr)Cl
C7H5BrCl2
MFCD00236034
2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
2-(bromomethyl)-1,4-dichlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents