Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzoic acid, 98+%

$75.28 - $854.30

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorobenzoic acid
CAS 50-79-3
Molecular Formula C7H4Cl2O2
Formula Weight 191.01
MDL Number MFCD00002416
Beilstein 973353
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1610814

  SDS
alfa aesar™
A1610814
25g Each for $75.28

AAA1610822

  SDS
alfa aesar™
A1610822
100g Each for $212.42

AAA1610836

  SDS
alfa aesar™
A1610836
500g Each for $854.30
Description & Specifications

Specifications

Formula Weight 191.01
Chemical Name or Material 2,5-Dichlorobenzoic acid
TSCA TSCA
Assay ≥98%
Molecular Formula C7H4Cl2O2
MDL Number MFCD00002416
CAS 50-79-3
Boiling Point 300° to 302°C
Beilstein 973353
Melting Point 151° to 154°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DG6825000
Quantity 25g

Applications
2,5-Dichlorobenzoic acid was used as calibration standard to investigate the gas-phase OH reaction products of biphenyl, monochlorobiphenyl and dichlorophenyl

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.