Learn More
2,5-Dichlorobenzoic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC163240100
Specifications
2, 5-Dichlorobenzoic acid | |
Glass Bottle | |
White to Beige | |
10g | |
50-79-3 | |
100.0 | |
C7H4Cl2O2 | |
MFCD00002416 | |
2,5-dichloro-benzoic acid, 2,5-dichlorobenzoicacid, 2,5dichlorobenzoic acid, benzoic acid, 2,5-dichloro, ccris 8609, dsstox_cid_4978, kp7cmr0gsd, pubchem3685, unii-kp7cmr0gsd, wln: qvr bg eg | |
QVTQYSFCFOGITD-UHFFFAOYSA-N | |
2,5-dichlorobenzoic acid | |
5784 | |
Powder |
Authentic | |
301.0°C | |
152.0°C to 157.0°C | |
97% | |
96.0 | |
96% min. (HPLC) | |
Cl2C6H3CO2H | |
09, 342 | |
Solubility in water: 0.8g/L (14°C). Other solubilities: soluble in most polar solvents,insoluble in non-polar solvents | |
C1=CC(=C(C=C1Cl)C(=O)O)Cl | |
191.01 | |
191.01 | |
97% |
Chemical Identifiers
50-79-3 | |
191.01 | |
QVTQYSFCFOGITD-UHFFFAOYSA-N | |
5784 | |
C1=CC(=C(C=C1Cl)C(=O)O)Cl |
C7H4Cl2O2 | |
MFCD00002416 | |
2,5-dichloro-benzoic acid, 2,5-dichlorobenzoicacid, 2,5dichlorobenzoic acid, benzoic acid, 2,5-dichloro, ccris 8609, dsstox_cid_4978, kp7cmr0gsd, pubchem3685, unii-kp7cmr0gsd, wln: qvr bg eg | |
2,5-dichlorobenzoic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-065-7
RTECSNumber : DG6825000
TSCA : TSCA