Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzhydrazide, 98+%

$86.19 - $298.34

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorobenzhydrazide
CAS 67487-35-8
Molecular Formula C7H6Cl2N2O
Formula Weight 205.04
MDL Number MFCD00014754
Beilstein 4394418
Synonym 2,5-Dichlorobenzoic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1477806

  SDS
alfa aesar™
A1477806
5g Each for $86.19

AAA1477814

  SDS
alfa aesar™
A1477814
25g Each for $298.34
Description & Specifications

Specifications

Formula Weight 205.04
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,5-Dichlorobenzhydrazide
MDL Number MFCD00014754
Synonym 2,5-Dichlorobenzoic acid hydrazide
Quantity 5g
Beilstein 4394418
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H6Cl2N2O
Assay ≥98%
Health Hazard 1 Warning
CAS 67487-35-8
Melting Point 178° to 181°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.