missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzhydrazide, 98+%

Manufacturer:  Alfa Aesar™ A1477814

 View more versions of this product

Catalog No. AAA1477814



Specifications

Specifications

2,5-Dichlorobenzhydrazide
25g
C7H6Cl2N2O
4394418
XECBCXNWGBDIMG-UHFFFAOYSA-N
2,5-dichlorobenzohydrazide
144246
≥98%
178°C to 181°C
67487-35-8
MFCD00014754
2,5-dichlorobenzhydrazide, 1,4-dichlorobenzene-2-carbohydrazide, #, 2,5-dichlorobenzoic hydrazide, 2,5-bis chloranyl benzohydrazide, labotest-bb lt00454263, labotest-bb lt03497761, 2,5-dichlorobenzoic acid hydrazide
C1=CC(=C(C=C1Cl)C(=O)NN)Cl
205.038
205.04

Chemical Identifiers

67487-35-8
205.038
XECBCXNWGBDIMG-UHFFFAOYSA-N
144246
C1=CC(=C(C=C1Cl)C(=O)NN)Cl
C7H6Cl2N2O
MFCD00014754
2,5-dichlorobenzhydrazide, 1,4-dichlorobenzene-2-carbohydrazide, #, 2,5-dichlorobenzoic hydrazide, 2,5-bis chloranyl benzohydrazide, labotest-bb lt00454263, labotest-bb lt03497761, 2,5-dichlorobenzoic acid hydrazide
2,5-dichlorobenzohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents