Please login to your online account to display your discounted pricing

2,5-Dichlorobenzenethiol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dichlorobenzenethiol
Name Note 98%
CAS 5858-18-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4Cl2S
Linear Formula Cl2C6H3SH
Formula Weight 179.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC162860250

  SDS
thermo fisher scientific
AC162860250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 179.07
Physical Form Liquid After Melting
Quantity 25g
Color Clear colorless to yellow
Beilstein 06, II, 298
CAS Max % 100.0
Molecular Formula C6H4Cl2S
MDL Number MFCD00004831
Chemical Name or Material 2, 5-Dichlorobenzenethiol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
CAS 5858-18-4
Name Note 98%
Assay Percent Range 98%
Linear Formula Cl2C6H3SH
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.