missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1480536

 View more versions of this product

Catalog No. AAA1480536



Specifications

Specifications

2,5-Dichlorobenzenesulfonyl chloride
36°C to 39°C
5402-73-3
MFCD00007429
2369410
BXCOSWRSIISQSL-UHFFFAOYSA-N
2,5-dichlorobenzenesulfonyl chloride
79401
98%
>110°C (230°F)
500g
C6H3Cl3O2S
UN3261
2,5-dichlorobenzene-1-sulfonyl chloride, 2,5-dichlorobenzenesulphonyl chloride, 2,5-dichlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2,5-dichloro, 2,5-dichloro-benzenesulfonyl chloride, 2,5-dichlorophenylsulfonyl chloride, 2,5-dichlorophenyl chlorosulfone, pubchem5116, acmc-209lcc, bxcoswrsiisqsl-uhfffaoysa
C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)Cl
245.498
245.51

Chemical Identifiers

5402-73-3
245.498
BXCOSWRSIISQSL-UHFFFAOYSA-N
79401
C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)Cl
C6H3Cl3O2S
MFCD00007429
2,5-dichlorobenzene-1-sulfonyl chloride, 2,5-dichlorobenzenesulphonyl chloride, 2,5-dichlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2,5-dichloro, 2,5-dichloro-benzenesulfonyl chloride, 2,5-dichlorophenylsulfonyl chloride, 2,5-dichlorophenyl chlorosulfone, pubchem5116, acmc-209lcc, bxcoswrsiisqsl-uhfffaoysa
2,5-dichlorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents