Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzenesulfonic acid dihydrate, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorobenzenesulfonic acid dihydrate
CAS 38484-94-5
Molecular Formula C6H4Cl2O3S·2H2O
Formula Weight 263.09 (227.07 Anhydrous)
MDL Number MFCD08271971
UN Number UN2585
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2565014

  SDS
alfa aesar™
B2565014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2565022

  SDS
alfa aesar™
B2565022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.09 (227.07 Anhydrous)
Health Hazard 1 Danger
Quantity 25g
Melting Point 98° to 102°C
Assay 97%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
Molecular Formula C6H4Cl2O3S·2H2O
Chemical Name or Material 2,5-Dichlorobenzenesulfonic acid dihydrate
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
CAS 38484-94-5
MDL Number MFCD08271971
UN Number UN2585
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.