Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorobenzamide, 97%

$62.34 - $220.12

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorobenzamide
CAS 5980-26-7
Molecular Formula C7H5Cl2NO
Formula Weight 190.03
MDL Number MFCD00017609
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2509706

 
alfa aesar™
B2509706
5g Each for $62.34

AAB2509714

 
alfa aesar™
B2509714
25g Each for $220.12
Description & Specifications

Specifications

Formula Weight 190.03
Chemical Name or Material 2,5-Dichlorobenzamide
Assay 97%
Molecular Formula C7H5Cl2NO
MDL Number MFCD00017609
CAS 5980-26-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 161°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.