missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichlorobenzaldehyde, 98%, ACROS Organics™

Chemical Identifiers

CAS 6361-23-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H4Cl2O
Linear Formula ClC6H3(Cl)CHO
Formula Weight 175.02
MDL Number MFCD00156140
Chemical Name or Material 2, 5-Dichlorobenzaldehyde, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC359940010 SDS Thermo Fisher Scientific
35994-0010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359940050 SDS Thermo Fisher Scientific
35994-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6361-23-5
100.0
C7H4Cl2O
175.02
2, 5-Dichlorobenzaldehyde, 98%
97.5
98%
ClC6H3(Cl)CHO
MFCD00156140
≥97.5%

Specifications

Crystalline Powder
1g
54° to 58°C
97.5
Authentic
98%
C7H4Cl2O
2, 5-Dichlorobenzaldehyde, 98%
175.02
White to off-white
100.0
Glass bottle
ClC6H3(Cl)CHO
MFCD00156140
6361-23-5
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 228-832-1

TSCA : TSCA