missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichloroanisole, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 264180250

 View more versions of this product

Catalog No. AC264180250



Specifications

Specifications

2, 5-Dichloroanisole
Authentic
1.5615 to 1.5635
Colorless to Yellow
99%
98.5
98.5% min. (GC)
Cl2C6H3OCH3
2,5-dichloroanisole, 2,5-dichloroanizole, 2,5-dichloromethoxybenzene, anisole, 2,5-dichloro, banair, benzene, 1,4-dichloro-2-methoxy, dsstox_cid_7452, pubchem3681, unii-8b3j0y3cj1
COC1=C(C=CC(=C1)Cl)Cl
177.03
177.03
99%
1.3300g/mL
Glass bottle
1.33
25g
1984-58-3
100.0
C7H6Cl2O
MFCD00061123
QKMNFFSBZRGHDJ-UHFFFAOYSA-N
1,4-dichloro-2-methoxybenzene
16125
Liquid

Chemical Identifiers

1984-58-3
177.03
QKMNFFSBZRGHDJ-UHFFFAOYSA-N
16125
COC1=C(C=CC(=C1)Cl)Cl
C7H6Cl2O
MFCD00061123
2,5-dichloroanisole, 2,5-dichloroanizole, 2,5-dichloromethoxybenzene, anisole, 2,5-dichloro, banair, benzene, 1,4-dichloro-2-methoxy, dsstox_cid_7452, pubchem3681, unii-8b3j0y3cj1
1,4-dichloro-2-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-852-6

TSCA : TSCA

SDS Documents