Please login to your online account to display your discounted pricing

2,5-Dichloroaniline 99%, Acros Organics


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146612500

 View more versions of this product

Catalog No. AC146612500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 162.02
Physical Form Crystalline Flakes
Quantity 250g
Color Beige to dark brown
Beilstein 12, IV, 1250
TSCA TSCA
EINECS Number 202-455-2
RTECS Number BX2610000
Melting Point 48° to 51°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Storage Information Product darkens on storage
Infrared Spectrum Authentic
Synonym C.I. 37010
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Molecular Formula C6H5Cl2N
MDL Number MFCD00007667
Name Note 99%
CAS 95-82-9
Chemical Name or Material 2, 5-Dichloroaniline
Assay Percent Range 99%
Linear Formula Cl2C6H3NH2

Specifications

Chemical Name or Material 2, 5-Dichloroaniline
Name Note 99%
CAS 95-82-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Formula Weight 162.02
MDL Number MFCD00007667
Synonym C.I. 37010
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents