Please login to your online account to display your discounted pricing

2,5-Dichloroaniline 99%, Acros Organics

$49.18 - $49.18

Chemical Identifiers

Chemical Name or Material 2, 5-Dichloroaniline
Name Note 99%
CAS 95-82-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Formula Weight 162.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146610010

  SDS
thermo fisher scientific
146610010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC146612500

  SDS
thermo fisher scientific
146612500
Glass bottle 250g Each for $49.18
Description & Specifications

Specifications

Physical Form Crystalline Flakes
Assay ≥98.5%
Formula Weight 162.02
Quantity 1kg
Color Beige to dark brown
EINECS Number 202-455-2
TSCA TSCA
Melting Point 48° to 51°C
RTECS Number BX2610000
Beilstein 12, IV, 1250
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
CAS Min % 98.5
Chemical Name or Material 2, 5-Dichloroaniline
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H5Cl2N
Storage Information Product darkens on storage
Linear Formula Cl2C6H3NH2
MDL Number MFCD00007667
CAS 95-82-9
Synonym C.I. 37010
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.