Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloroaniline, 99%

$79.41 - $79.41

Chemical Identifiers

Chemical Name or Material 2,5-Dichloroaniline
CAS 95-82-9
Molecular Formula C6H5Cl2N
Formula Weight 162.02
MDL Number MFCD00007667
UN Number UN3442
Beilstein 1447438
Synonym C.I. 37010
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1115130

  SDS
alfa aesar™
A1115130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1115136

  SDS
alfa aesar™
A1115136
500g Each for $79.41

AAA111510E

  SDS
alfa aesar™
A111510E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,5-Dichloroaniline
Flash Point 139°C (282°F)
Boiling Point 250° to 251°C
Melting Point 48° to 52°C
Quantity 250g
CAS 95-82-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
RTECS Number BX2610000
TSCA TSCA
Assay 99%
Molecular Formula C6H5Cl2N
Formula Weight 162.02
MDL Number MFCD00007667
UN Number UN3442
Beilstein 1447438
Synonym C.I. 37010

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.