missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloroaniline, 99%

Manufacturer:  Alfa Aesar™ A1115130

 View more versions of this product

Catalog No. AAA1115130


 

Description

Description

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,5-Dichloroaniline
250°C to 251°C
250g
99%
MFCD00007667
1447438
AVYGCQXNNJPXSS-UHFFFAOYSA-N
2,5-dichloroaniline
7262
162.02
139°C (282°F)
48°C to 52°C
95-82-9
C6H5Cl2N
UN3442
C.I. 37010
C1=CC(=C(C=C1Cl)N)Cl
162.013
CHEBI:34245

Chemical Identifiers

95-82-9
162.013
AVYGCQXNNJPXSS-UHFFFAOYSA-N
7262
C6H5Cl2N
MFCD00007667
C.I. 37010
CHEBI:34245
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID

RTECSNumber : BX2610000

TSCA : TSCA

SDS Documents