Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Dichloroacetophenone, 98%

$100.96 - $417.38

Chemical Identifiers

Chemical Name or Material 2',5'-Dichloroacetophenone
CAS 2476-37-1
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00000607
Beilstein 1865043
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1377014

  SDS
alfa aesar™
A1377014
25g Each for $100.96

AAA1377022

  SDS
alfa aesar™
A1377022
100g Each for $417.38

AAA1377009

  SDS
alfa aesar™
A1377009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2',5'-Dichloroacetophenone
Quantity 25g
CAS 2476-37-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA
Assay 98%
Molecular Formula C8H6Cl2O
Formula Weight 189.04
MDL Number MFCD00000607
Boiling Point 118°C (12mmHg)
Beilstein 1865043
Melting Point 11° to 13°C
Flash Point >110°C (230°F)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.