Please login to your online account to display your discounted pricing

2',5'-Dichloroacetoacetanilide, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2', 5'-Dichloroacetoacetanilide
Assay Percent Range 98%
Molecular Formula C10H9Cl2NO2
Formula Weight 246.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC406710010

  SDS
thermo fisher scientific
AC406710010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC406710050

  SDS
thermo fisher scientific
AC406710050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.09
Quantity 1g
CAS 2044-72-6
Chemical Name or Material 2', 5'-Dichloroacetoacetanilide
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H9Cl2NO2