Please login to your online account to display your discounted pricing

2',5'-Dichloroacetoacetanilide, 98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC406710050

 View more versions of this product

Catalog No. AC406710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 246.09
Quantity 5g
Chemical Name or Material 2', 5'-Dichloroacetoacetanilide
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H9Cl2NO2
CAS 2044-72-6

Specifications

Chemical Name or Material 2', 5'-Dichloroacetoacetanilide
Assay Percent Range 98%
Molecular Formula C10H9Cl2NO2
Formula Weight 246.09