Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Dichloroacetoacetanilide, 98%

$35.09 - $136.23

Chemical Identifiers

Chemical Name or Material 2',5'-Dichloroacetoacetanilide
CAS 2044-72-6
Molecular Formula C10H9Cl2NO2
Formula Weight 246.09
MDL Number MFCD00018520
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1670006

 
alfa aesar™
A1670006
5g Each for $35.09

AAA1670014

 
alfa aesar™
A1670014
25g Each for $136.23
Description & Specifications

Specifications

Formula Weight 246.09
Chemical Name or Material 2',5'-Dichloroacetoacetanilide
TSCA TSCA
Assay 98%
Molecular Formula C10H9Cl2NO2
MDL Number MFCD00018520
CAS 2044-72-6
Melting Point 94° to 96°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.