Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-D-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dichloro-D-phenylalanine
CAS 718596-54-4
Formula Weight 234.08
MDL Number MFCD07372016
Synonym H-D-Phe(2,5-DiCl)-OH; (R)-2-Amino-3-(2,5-dichlorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6677303

 
alfa aesar™
H6677303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66773MD

 
alfa aesar™
H66773MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.08
Chemical Name or Material 2,5-Dichloro-D-phenylalanine
MDL Number MFCD07372016
CAS 718596-54-4
Assay 98%
Synonym H-D-Phe(2,5-DiCl)-OH; (R)-2-Amino-3-(2,5-dichlorophenyl)propionic acid
Physical Form Solid
Quantity 1g