Please login to your online account to display your discounted pricing

2,5-Dichloro-4-phenylenediamine, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 5-Dichloro-4-phenylenediamine
CAS 20103-09-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H2(NH2)2
Formula Weight 177.03
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC199090250

  SDS
thermo fisher scientific
199090250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC199090050

  SDS
thermo fisher scientific
199090050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 177.03
Color Light brown to pinkish-gray
Quantity 25g
Beilstein 13, 118
EINECS Number 243-512-1
TSCA TSCA
RTECS Number SS9150000
Melting Point 164° to 168°C
CAS Max % 100.0
Assay Percent Range 98%
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 5-Dichloro-4-phenylenediamine
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H2(NH2)2
CAS 20103-09-7
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.