Please login to your online account to display your discounted pricing

2,5-Dichloro-3-nitropyridine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dichloro-3-nitropyridine
CAS 21427-62-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H2Cl2N2O2
Formula Weight 192.99
MDL Number MFCD06658963
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379520010

  SDS
fisher scientific acros
AC379520010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379520050

  SDS
fisher scientific acros
AC379520050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 192.99
Physical Form Crystalline Powder and/or Chunks
Quantity 1g
Color Light beige-green to orange
Melting Point 39° to 45°C
Beilstein 20, IV, 2532
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD06658963
Chemical Name or Material 2, 5-Dichloro-3-nitropyridine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 21427-62-3
Packaging Glass bottle
Molecular Formula C5H2Cl2N2O2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.