missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-3-nitropyridine, ≥97%

Manufacturer:  Alfa Aesar™ H6427606

 View more versions of this product

Catalog No. AAH6427606



Specifications

Specifications

2,5-Dichloro-3-nitropyridine
5g
C5H2Cl2N2O2
UN2811
2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
192.983
192.98
≥97%
41°C to 45°C
21427-62-3
MFCD06658963
147539
OBUGJYJQJWMOQO-UHFFFAOYSA-N
2,5-dichloro-3-nitropyridine
8063331
Solid

Chemical Identifiers

21427-62-3
192.983
OBUGJYJQJWMOQO-UHFFFAOYSA-N
8063331
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
C5H2Cl2N2O2
MFCD06658963
2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
2,5-dichloro-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents