missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-3-nitrobenzoic acid, 98+%

$52.58 - $161.92

Chemical Identifiers

CAS 88-86-8
Molecular Formula C7H3Cl2NO4
Molecular Weight (g/mol) 236.004
MDL Number MFCD00014691
InChI Key AUAXYMOBWXOEQD-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-nitrobenzoic acid, dinoben, benzoic acid, 2,5-dichloro-3-nitro, caswell no. 312, 3-nitro-2,5-dichlorobenzoic acid, unii-5rxk12d272, kyselina 2,5-dichlor-3-nitrobenzoova, epa pesticide chemical code 028101, kyselina 2,5-dichlor-3-nitrobenzoova czech, 2,5-dichloro-3-nitro-benzoic acid
PubChem CID 6951
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0133203 View Documents Alfa Aesar™
L0133203
1g Each for $52.58
 
AAL0133206 View Documents Alfa Aesar™
L0133206
5g Each for $161.92
 
Specifications

Chemical Identifiers

88-86-8
236.004
AUAXYMOBWXOEQD-UHFFFAOYSA-N
6951
C7H3Cl2NO4
MFCD00014691
2,5-dichloro-3-nitrobenzoic acid, dinoben, benzoic acid, 2,5-dichloro-3-nitro, caswell no. 312, 3-nitro-2,5-dichlorobenzoic acid, unii-5rxk12d272, kyselina 2,5-dichlor-3-nitrobenzoova, epa pesticide chemical code 028101, kyselina 2,5-dichlor-3-nitrobenzoova czech, 2,5-dichloro-3-nitro-benzoic acid
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)Cl

Specifications

218°C to 222°C
88-86-8
MFCD00014691
2,5-dichloro-3-nitrobenzoic acid, dinoben, benzoic acid, 2,5-dichloro-3-nitro, caswell no. 312, 3-nitro-2,5-dichlorobenzoic acid, unii-5rxk12d272, kyselina 2,5-dichlor-3-nitrobenzoova, epa pesticide chemical code 028101, kyselina 2,5-dichlor-3-nitrobenzoova czech, 2,5-dichloro-3-nitro-benzoic acid
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)C(=O)O)Cl
6951
≥98%
1g
C7H3Cl2NO4
1976119
AUAXYMOBWXOEQD-UHFFFAOYSA-N
236.004
236.01
2,5-Dichloro-3-nitrobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG7875000