missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichloro-3-methylthiophene 96.0+%, TCI America™

$55.97 - $179.70

Chemical Identifiers

CAS 17249-90-0
Molecular Formula C5H4Cl2S
Molecular Weight (g/mol) 167.05
MDL Number MFCD00022490
InChI Key BUWSFDLUTRHPBO-UHFFFAOYSA-N
Synonym 2,5-dichlor-3-methylthiophen, 2,5-dichloro-3-methyl thiophene, 2,5-dichloro-3-methyl-thiophene, 2,5-dichloro-3-methylthiophene, acmc-209e4q, pubchem5195
PubChem CID 268910
IUPAC Name 2,5-dichloro-3-methylthiophene
SMILES CC1=C(Cl)SC(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D36801G
View Documents
Tci America
D36801G
1g
Each for $55.97
 
D36805G
View Documents
Tci America
D36805G
5g
Each for $179.70
 
Description

Description

Specifications

Chemical Identifiers

17249-90-0
167.05
BUWSFDLUTRHPBO-UHFFFAOYSA-N
268910
CC1=C(Cl)SC(Cl)=C1
C5H4Cl2S
MFCD00022490
2,5-dichlor-3-methylthiophen, 2,5-dichloro-3-methyl thiophene, 2,5-dichloro-3-methyl-thiophene, 2,5-dichloro-3-methylthiophene, acmc-209e4q, pubchem5195
2,5-dichloro-3-methylthiophene

Specifications

185°C
1g
C5H4Cl2S
2,5-dichlor-3-methylthiophen, 2,5-dichloro-3-methyl thiophene, 2,5-dichloro-3-methyl-thiophene, 2,5-dichloro-3-methylthiophene, acmc-209e4q, pubchem5195
CC1=C(Cl)SC(Cl)=C1
167.05
167.05
≥96.0% (GC)
Yellow
17249-90-0
MFCD00022490
BUWSFDLUTRHPBO-UHFFFAOYSA-N
2,5-dichloro-3-methylthiophene
268910
Clear Liquid at 20°C
2,5-Dichloro-3-methylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS