Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-3-iodopyridine, 98%

$120.38 - $270.92

Chemical Identifiers

Chemical Name or Material 2,5-Dichloro-3-iodopyridine
CAS 942206-23-7
Molecular Formula C5H2Cl2IN
Formula Weight 273.89
MDL Number MFCD09037444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2778603

 
alfa aesar™
H2778603
1g Each for $270.92

AAH27786MD

 
alfa aesar™
H27786MD
250mg Each for $120.38
Description & Specifications

Specifications

Formula Weight 273.89
Chemical Name or Material 2,5-Dichloro-3-iodopyridine
Assay 98%
Molecular Formula C5H2Cl2IN
MDL Number MFCD09037444
CAS 942206-23-7
Melting Point 51° to 53°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.