Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-3,4-dinitrothiophene, 97+%

$57.09 - $227.81

Chemical Identifiers

Chemical Name or Material 2,5-Dichloro-3,4-dinitrothiophene
CAS 51584-21-5
Molecular Formula C4Cl2N2O4S
Formula Weight 243.03
MDL Number MFCD00052395
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1127803

 
alfa aesar™
A1127803
1g Each for $57.09

AAA1127806

 
alfa aesar™
A1127806
5g Each for $227.81
Description & Specifications

Specifications

Formula Weight 243.03
Chemical Name or Material 2,5-Dichloro-3,4-dinitrothiophene
Assay ≥97%
Molecular Formula C4Cl2N2O4S
MDL Number MFCD00052395
CAS 51584-21-5
Melting Point 85° to 89°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.