Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloro-1,4-benzoquinone, 98%

$40.16 - $181.64

Chemical Identifiers

Chemical Name or Material 2,5-Dichloro-1,4-benzoquinone
CAS 615-93-0
Molecular Formula C6H2Cl2O2
Formula Weight 176.98
MDL Number MFCD00041736
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2715203

  SDS
alfa aesar™
H2715203
1g Each for $40.16

AAH2715209

  SDS
alfa aesar™
H2715209
10g Each for $181.64
Description & Specifications

Specifications

Formula Weight 176.98
Chemical Name or Material 2,5-Dichloro-1,4-benzoquinone
TSCA TSCA
Assay 98%
Molecular Formula C6H2Cl2O2
MDL Number MFCD00041736
CAS 615-93-0
Melting Point 158° to 162°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DK3990000
Quantity 1g

Applications
2,5-Dichloro-1,4-benzoquinone, is used as a agrochemical, pharmaceutical intermediate. It is also used in the organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.