missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dibromotoluene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 263480050

 View more versions of this product

Catalog No. AC263480050



Description

Description

Specifications

Specifications

2, 5-Dibromotoluene
>110°C
Glass bottle
1.81
Colorless to Yellow
5g
615-59-8
100.0
C7H6Br2
MFCD00000090
1,4-dibromo-2-methyl-benzene, 2,5-dibromo toluene, 2,5-dibromotoluene, 3,6-dibromotoluene, acmc-209mu2, benzene, 1,4-dibromo-2-methyl, chemwish ic06904, pubchem3922, toluene, 2,5-dibromo
QKEZTJYRBHOKHH-UHFFFAOYSA-N
1,4-dibromo-2-methylbenzene
12006
Liquid
1.8100g/mL
Authentic
1.6010 to 1.6030
135.0°C to 136.0°C (35.0mmHg)
5.0°C to 6.0°C
99%
98.5
97.5% min. (GC)
CH3C6H3Br2
05, 308
Solubility in water: immiscible
CC1=C(C=CC(=C1)Br)Br
249.93
249.93
98%

Chemical Identifiers

615-59-8
249.93
QKEZTJYRBHOKHH-UHFFFAOYSA-N
12006
CC1=C(C=CC(=C1)Br)Br
C7H6Br2
MFCD00000090
1,4-dibromo-2-methyl-benzene, 2,5-dibromo toluene, 2,5-dibromotoluene, 3,6-dibromotoluene, acmc-209mu2, benzene, 1,4-dibromo-2-methyl, chemwish ic06904, pubchem3922, toluene, 2,5-dibromo
1,4-dibromo-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 210-437-0

TSCA : TSCA

SDS Documents