missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dibromotoluene, 99%, ACROS Organics™

$31.14 - $139.89

Chemical Identifiers

CAS 615-59-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H6Br2
Linear Formula CH3C6H3Br2
Formula Weight 249.93
MDL Number MFCD00000090
Chemical Name or Material 2, 5-Dibromotoluene, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC263480050 SDS Thermo Fisher Scientific
263480050
Glass bottle 5g Each for $31.14
AC263480250 SDS Thermo Fisher Scientific
263480250
Glass bottle 25g Each for $139.89
Specifications Description & Specifications

Chemical Identifiers

615-59-8
100.0
C7H6Br2
249.93
2, 5-Dibromotoluene, 99%
98.5
99%
CH3C6H3Br2
MFCD00000090
≥98.5%

Specifications

615-59-8
100.0
Glass bottle
Liquid
C7H6Br2
249.93
05, 308
Clear colorless to yellow
2, 5-Dibromotoluene, 99%
98.5
Authentic
1.6010 to 1.6030 (20°C, 589nm)
99%
CH3C6H3Br2
MFCD00000090
1.8100g/mL
5g
≥98.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-437-0

TSCA : TSCA