Please login to your online account to display your discounted pricing

2,5-Dibromoterephthalic acid, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB11564DE

 View more versions of this product

Catalog No. BTB11564DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 323.92
Quantity 5g
Assay 97%
Molecular Formula C8H4Br2O4
Linear Formula Br2C6H2(CO2H)2
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 13731-82-3
Chemical Name or Material 2, 5-Dibromoterephthalic acid

Specifications

Chemical Name or Material 2, 5-Dibromoterephthalic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H4Br2O4
Linear Formula Br2C6H2(CO2H)2
Formula Weight 323.92