Please login to your online account to display your discounted pricing

2,5-Dibromoterephthalic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 5-Dibromoterephthalic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H4Br2O4
Linear Formula Br2C6H2(CO2H)2
Formula Weight 323.92
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB11564DA

  SDS
fisher scientific company
BTB11564DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 323.92
Quantity 1g
Assay 97%
Chemical Name or Material 2, 5-Dibromoterephthalic acid
Molecular Formula C8H4Br2O4
Packaging Amber Glass Bottle
Linear Formula Br2C6H2(CO2H)2
CAS 13731-82-3
Assay Percent Range 97%