missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromohydroquinone, 97%

Manufacturer:  Alfa Aesar™ H5545314

 View more versions of this product

Catalog No. AAH5545314



Description

Description

Applications
It is an important pharmaceutical intermediate.

Solubility
Soluble in methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications

Specifications

2,5-Dibromohydroquinone
25g
C6H4Br2O2
1,4-benzenediol, 2,5-dibromo
VALXCIRMSIFPFN-UHFFFAOYSA-N
2,5-dibromobenzene-1,4-diol
280945
97%
191°C to 194°C
14753-51-6
MFCD00192664
Soluble in methanol
C1=C(C(=CC(=C1Br)O)Br)O
267.904
267.9

Chemical Identifiers

14753-51-6
267.904
VALXCIRMSIFPFN-UHFFFAOYSA-N
280945
C1=C(C(=CC(=C1Br)O)Br)O
C6H4Br2O2
MFCD00192664
1,4-benzenediol, 2,5-dibromo
2,5-dibromobenzene-1,4-diol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents