Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromohexane, erythro + threo, 97%

$64.03 - $237.05

Chemical Identifiers

Chemical Name or Material 2,5-Dibromohexane
CAS 24774-58-1
Name Note erythro + threo
Molecular Formula C6H12Br2
Formula Weight 243.98
MDL Number MFCD00039177
Beilstein 1732460
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1258106

 
alfa aesar™
L1258106
5g Each for $64.03

AAL1258114

 
alfa aesar™
L1258114
25g Each for $237.05
Description & Specifications

Specifications

Formula Weight 243.98
Chemical Name or Material 2,5-Dibromohexane
TSCA TSCA
Boiling Point 88° to 89°C (13mmHg)
Assay 97%
Molecular Formula C6H12Br2
MDL Number MFCD00039177
CAS 24774-58-1
Beilstein 1732460
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Name Note erythro + threo
Quantity 5g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.