Please login to your online account to display your discounted pricing

2,5-Dibromohexane 97%, mixture of diastereomers, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC389130050

 View more versions of this product

Catalog No. AC389130050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (mixed diastereosomers)
Formula Weight 243.97
Physical Form Liquid
Quantity 5g
Color Clear colorless to pale yellow
CAS Min % 96.0
CAS Max % 100.0
Linear Formula BrCH(CH3)CH2CH2CH(CH3)Br
Packaging Glass bottle
Refractive Index 1.5000 to 1.5020 (20°C, 589nm)
MDL Number MFCD00039177
CAS 24774-58-1
Chemical Name or Material 2, 5-Dibromohexane
Infrared Spectrum Authentic
Molecular Formula C6 H12 Br2
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 5-Dibromohexane
CAS 24774-58-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (mixed diastereosomers)
Assay Percent Range 97%
Molecular Formula C6 H12 Br2
Linear Formula BrCH(CH3)CH2CH2CH(CH3)Br
Formula Weight 243.97
MDL Number MFCD00039177
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents