Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromobenzotrifluoride, 98%

$97.95 - $364.64

Chemical Identifiers

Chemical Name or Material 2,5-Dibromobenzotrifluoride
CAS 7657-09-2
Molecular Formula C7H3Br2F3
Formula Weight 303.91
MDL Number MFCD00013554
Beilstein 2261093
Synonym 1,4-Dibromo-2-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0337806

  SDS
alfa aesar™
L0337806
5g Each for $97.95

AAL0337814

  SDS
alfa aesar™
L0337814
25g Each for $364.64
Description & Specifications

Specifications

Formula Weight 303.91
Chemical Name or Material 2,5-Dibromobenzotrifluoride
CAS 7657-09-2
Health Hazard 1 Warning
Beilstein 2261093
Assay 98%
Melting Point 47° to 51°C
MDL Number MFCD00013554
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C7H3Br2F3
Flash Point >100°C (212°F)
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 1,4-Dibromo-2-(trifluoromethyl)benzene
Boiling Point 210°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.