missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1507714

 View more versions of this product

Catalog No. AAA1507714



Specifications

Specifications

2,5-Dibromobenzoic acid
25g
C7H4Br2O2
1868193
SQQKOTVDGCJJKI-UHFFFAOYSA-N
2,5-dibromobenzoic acid
11891
98%
153°C to 157°C
610-71-9
MFCD00016494
benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
C1=CC(=C(C=C1Br)C(=O)O)Br
279.915
279.93

Chemical Identifiers

610-71-9
279.915
SQQKOTVDGCJJKI-UHFFFAOYSA-N
11891
C1=CC(=C(C=C1Br)C(=O)O)Br
C7H4Br2O2
MFCD00016494
benzoic acid, 2,5-dibromo, rarechem fh 1w 0049, pubchem3859, 2,5-dibromobenzoicacid, 2,5-dibromo-benzoic acid, benzoicacid, 2,5-dibromo, 4-09-00-01027 beilstein handbook reference, ksc353s4r, acmc-1b114, 2,5-dibromobenzoic acid
2,5-dibromobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG6290000

Documents