Learn More
2,5-Dibromoaniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 155030050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 5-Dibromoaniline | |
3638-73-1 | |
100.0 | |
Authentic | |
Glass bottle | |
Br2C6H3NH2 | |
5g | |
2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline | |
C1=CC(=C(C=C1Br)N)Br | |
250.92 | |
250.92 |
97% | |
96.0 | |
109°C | |
96% min. (GC) | |
C6H5Br2N | |
MFCD00007636 | |
12, 659 | |
WRTAZRGRFBCKBU-UHFFFAOYSA-N | |
2,5-dibromoaniline | |
77198 | |
97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolo
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POI
GHS Signal Word: Danger
EINECSNumber : 222-865-5
TSCA : TSCA
RUO â Research Use Only