missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide 98.0+%, TCI America™
$199.08 - $690.14
Chemical Identifiers
CAS | 1231160-83-0 |
---|---|
Molecular Formula | C14H17Br2NO2S |
Molecular Weight (g/mol) | 423.163 |
MDL Number | MFCD23703120 |
InChI Key | AOZLCBYWDXFKCJ-UHFFFAOYSA-N |
Synonym | 1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione |
PubChem CID | 58261078 |
IUPAC Name | 1,3-dibromo-5-(2-ethylhexyl)thieno[3,4-c]pyrrole-4,6-dione |
SMILES | CCCCC(CC)CN1C(=O)C2=C(SC(=C2C1=O)Br)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D5021200MG
|
TCI America
D5021200MG |
200 mg |
Each for $199.08
|
|
|||||
D50211G
|
TCI America
D50211G |
1 g |
Each for $690.14
|
|
|||||
Chemical Identifiers
1231160-83-0 | |
423.163 | |
AOZLCBYWDXFKCJ-UHFFFAOYSA-N | |
58261078 | |
CCCCC(CC)CN1C(=O)C2=C(SC(=C2C1=O)Br)Br |
C14H17Br2NO2S | |
MFCD23703120 | |
1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione | |
1,3-dibromo-5-(2-ethylhexyl)thieno[3,4-c]pyrrole-4,6-dione |
Specifications
1231160-83-0 | |
White-Yellow | |
MFCD23703120 | |
1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione | |
CCCCC(CC)CN1C(=O)C2=C(SC(=C2C1=O)Br)Br | |
423.163 | |
423.16 | |
Crystalline Powder |
112°C | |
C14H17Br2NO2S | |
200 mg | |
AOZLCBYWDXFKCJ-UHFFFAOYSA-N | |
1,3-dibromo-5-(2-ethylhexyl)thieno[3,4-c]pyrrole-4,6-dione | |
58261078 | |
≥98.0% (GC) | |
2,5-Dibromo-N-(2-ethylhexyl)-3,4-thiophenedicarboximide |
Safety and Handling
TSCA : No