Please login to your online account to display your discounted pricing

2,5-Dibromo-4-picoline, 98%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC361050250

 View more versions of this product

Catalog No. AC361050250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Assay ≥97.5%
Formula Weight 251
Quantity 25g
Color Light yellow to orange
Melting Point 37° to 41°C
CAS Max % 100.0
Molecular Formula C6H5Br2N
MDL Number MFCD00234955
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 5-Dibromo-4-picoline
CAS 3430-26-0

Specifications

Chemical Name or Material 2, 5-Dibromo-4-picoline
CAS 3430-26-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H5Br2N
Formula Weight 251
MDL Number MFCD00234955
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

SDS & Safety Documents