Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-4-methylpyridine, 97%

$294.00 - $294.00

Chemical Identifiers

Chemical Name or Material 2,5-Dibromo-4-methylpyridine
CAS 3430-26-0
Molecular Formula C6H5Br2N
Formula Weight 250.92
MDL Number MFCD00234955
Synonym 2,5-Dibromo-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3045706

  SDS
alfa aesar™
H3045706
5g Each for $294.00
Description & Specifications

Specifications

Formula Weight 250.92
Chemical Name or Material 2,5-Dibromo-4-methylpyridine
Melting Point 36° to 40°C
Assay 97%
Quantity 5g
Molecular Formula C6H5Br2N
MDL Number MFCD00234955
CAS 3430-26-0
Synonym 2,5-Dibromo-4-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.