Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-4-fluorotoluene, 96%

$108.34 - $363.14

Chemical Identifiers

Chemical Name or Material 2,5-Dibromo-4-fluorotoluene
CAS 134332-29-9
Molecular Formula C7H5Br2F
Formula Weight 267.92
MDL Number MFCD00044738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2429722

  SDS
alfa aesar™
B2429722
100g Each for $108.34

AAB2429736

  SDS
alfa aesar™
B2429736
500g Each for $363.14
Description & Specifications

Specifications

Formula Weight 267.92
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 2,5-Dibromo-4-fluorotoluene
Boiling Point 92° to 94°C (1mmHg)
Quantity 100g
CAS 134332-29-9
Molecular Formula C7H5Br2F
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
MDL Number MFCD00044738
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.