missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-nitropyridine, 98%

$57.78 - $876.64

Chemical Identifiers

CAS 15862-37-0
Molecular Formula C5H2Br2N2O2
Molecular Weight (g/mol) 281.891
MDL Number MFCD09266223
InChI Key OQKWPJCAKRVADO-UHFFFAOYSA-N
Synonym 2 pound not5-dibromo-3-nitropyridine, 2,5-di bromo-3-nitro pyridine, 2,5-dibromo-3-nitro-pyridine, acmc-1bp61, ksc495e2t, pubchem5505, pyridine, 2,5-dibromo-3-nitro
PubChem CID 298470
IUPAC Name 2,5-dibromo-3-nitropyridine
SMILES C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6196106
View Documents
Alfa Aesar™
H6196106
5g Each for $57.78
 
AAH6196114
View Documents
Alfa Aesar™
H6196114
25g Each for $244.61
 
AAH6196122
View Documents
Alfa Aesar™
H6196122
100g Each for $876.64
 
Specifications

Chemical Identifiers

15862-37-0
281.891
OQKWPJCAKRVADO-UHFFFAOYSA-N
298470
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
C5H2Br2N2O2
MFCD09266223
2 pound not5-dibromo-3-nitropyridine, 2,5-di bromo-3-nitro pyridine, 2,5-dibromo-3-nitro-pyridine, acmc-1bp61, ksc495e2t, pubchem5505, pyridine, 2,5-dibromo-3-nitro
2,5-dibromo-3-nitropyridine

Specifications

Yellow
15862-37-0
MFCD09266223
OQKWPJCAKRVADO-UHFFFAOYSA-N
2,5-dibromo-3-nitropyridine
298470
Solid
2,5-Dibromo-3-nitropyridine
5g
C5H2Br2N2O2
2 pound not5-dibromo-3-nitropyridine, 2,5-di bromo-3-nitro pyridine, 2,5-dibromo-3-nitro-pyridine, acmc-1bp61, ksc495e2t, pubchem5505, pyridine, 2,5-dibromo-3-nitro
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
281.891
281.89
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures