missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-nitropyridine, 98%

Manufacturer:  Alfa Aesar™ H6196114

 View more versions of this product

Catalog No. AAH6196114



Specifications

Specifications

2,5-Dibromo-3-nitropyridine
25g
C5H2Br2N2O2
2 pound not5-dibromo-3-nitropyridine, 2,5-di bromo-3-nitro pyridine, 2,5-dibromo-3-nitro-pyridine, acmc-1bp61, ksc495e2t, pubchem5505, pyridine, 2,5-dibromo-3-nitro
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
281.891
281.89
98%
Yellow
15862-37-0
MFCD09266223
OQKWPJCAKRVADO-UHFFFAOYSA-N
2,5-dibromo-3-nitropyridine
298470
Solid

Chemical Identifiers

15862-37-0
281.891
OQKWPJCAKRVADO-UHFFFAOYSA-N
298470
C1=C(C(=NC=C1Br)Br)[N+](=O)[O-]
C5H2Br2N2O2
MFCD09266223
2 pound not5-dibromo-3-nitropyridine, 2,5-di bromo-3-nitro pyridine, 2,5-dibromo-3-nitro-pyridine, acmc-1bp61, ksc495e2t, pubchem5505, pyridine, 2,5-dibromo-3-nitro
2,5-dibromo-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents