missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dibromo-3-n-octylthiophene, 96%

Manufacturer:  Alfa Aesar™ H5508206

 View more versions of this product

Catalog No. AAH5508206



Description

Description

Applications
It is a conducting polymer precursor.

Solubility
Slightly soluble in water.

Notes
Store at 2-8°C. Keep away from oxidizing agents.
Specifications

Specifications

2,5-Dibromo-3-n-octylthiophene
110°C (230°F)
316°C
149703-84-4
MFCD00672013
Slightly soluble in water.
CCCCCCCCC1=C(SC(=C1)Br)Br
354.144
354.14
1.44
1.548
5g
C12H18Br2S
2,5-bis bromanyl-3-octyl-thiophene
PJGBSUPPENVFAD-UHFFFAOYSA-N
2,5-dibromo-3-octylthiophene
4385790
96%

Chemical Identifiers

149703-84-4
354.144
PJGBSUPPENVFAD-UHFFFAOYSA-N
4385790
CCCCCCCCC1=C(SC(=C1)Br)Br
C12H18Br2S
MFCD00672013
2,5-bis bromanyl-3-octyl-thiophene
2,5-dibromo-3-octylthiophene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents